Huber Hotbox Series Heating Circulators
Designed specifically to provide heat to external open systems
  • Rugged heater, housing, and pump are constructed of stainless steel
  • Intuitive Pilot ONE touchscreen controller
  • Store and load temperature settings to and from a USB stick (not included)
The Hotbox flow-through heaters feature user-adjustable overtemperature protection, high-efficiency heating, intelligent energy management, and powerful circulation pump — ideal and reliable for continuous use. Developed exclusively for external open applications, there is no expansion tank. Fluid flows through the unit to be heated. The compact housing with castors allows these units to fit into any pilot plant.
Benefits of the integrated Pilot ONE controller include a large, 5.7" color, touchscreen display with 11 language support (EN, DE, FR, IT, ES, RU, CN, PT, JP, CZ, and PL), convenient color-coded navigation system, and USB and Ethernet ports for communication and remote control through your tablet, PC, or network.
$23,132.22CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Power (VAC) Description
Availability Pricing
Huber Hotbox 45 Heating Circulating Bath; 208VAC, 2 phase, 60 Hz RK-12125-78 Mfr # 2030.0002.01
COMPARE
45 208 Heating Circulating Bath; 208VAC, 2 phase, 60 Hz
$23,132.22CAD / Each