This product is now discontinued, View Related Products
Horiba Laqua F-70 Series pH/Ion, Conductivity and pH/Conductivity Meters
Intuitive touch-screen operation makes it easy to navigate
  • On-screen support for setup, maintenance, and troubleshooting
  • Auto hold function for calibration and measurement
  • Up to 2000 GLP/GMP data sets
  • USB PC communication and USB memory
The Laqua F-70 series of pH/Ion, Conductivity and pH/Conductivity meters are designed for simple, intuitive operation. These meters offer autocalibration, one-button measurement function, and easy-to-use touch-screen operation. Simply slide your finger across the screen to switch displays. The meter''s prompts provide support for meter setup, maintenance information, and troubleshooting tips to guide you through trouble-free operation.
Large color LCD displays measurement value and temperature. Units also feature Auto-hold to ensure stable readings. USB PC communication and USB memory for easy download of GLP/GMP data sets.
$997.12 - $3,502.25CAD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Horiba PH-1200 METERPH/MV/T BT METER RK-58702-00 Mfr # 3200647408
COMPARE
$997.12CAD / Each
Horiba Laqua F-72 Benchtop Single Channel pH/Ion Meter RK-58702-02 Mfr # 3200575120
COMPARE
Benchtop pH/Ion Meter
$2,098.11CAD / Each
Horiba Laqua F-73 Benchtop Dual Channel pH/Ion Meter RK-58702-04 Mfr # 3200575123
COMPARE
Benchtop Dual Channel pH/Ion Meter
$3,502.25CAD / Each
Horiba Laqua F-74 Benchtop Dual Channel pH/Ion/Conductivity/Resistivity/Salinity/TDS  Meter RK-58702-06 Mfr # 3200575130
COMPARE
Benchtop Dual Channel pH/Ion/Conductivity Meter
$3,282.71CAD / Each
Horiba Laqua EC-1100 Benchtop Conductivity/Resistivity/Salinity/TDS Meter RK-58702-10 Mfr # 3200647411
COMPARE
Benchtop Conductivity/Resistivity/Salinity/TDS Meter
$1,436.56CAD / Each
Horiba Laqua DS-72 Benchtop Conductivity/Resistivity/Salinity/TDS Meter RK-58702-12 Mfr # 3200575136
COMPARE
Benchtop Conductivity Meter
$2,173.25CAD / Each