This product is now discontinued, View Related Products
Honeywell Laser Eyewear; LOTG Series
Excellent visible light transmission minimizes distortion
These lightweight, impact-resistant polymer lenses are suitable for low-powered lasers or indirect laser exposure. Wavelength, optical density, and lot number are inscribed across lens for easy identification. All eyewear comes with protective case, cleaning cloth, and safety hanging cord.
LOTG Series
Eyewear features a broad, single-lens design to ensure maximum top and side protection. Glasses even fit comfortably over most prescription glasses.
Compliance: ANSI Z136.1-1986, Z87.1-2003 (glasses unsuitable for direct laser radiation).
$263.74 - $360.98CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Laser Type Lens Color
Availability Pricing
Honeywell LOTG-DIODE/YAG LOTG Laser Eyewear, Diode/YAG RK-86438-13 Mfr # LOTG-NDGAYAG
COMPARE
YAG, diode, CO2 Green
$263.74CAD / Each
Honeywell LOTG-YAG/CO2 Laser Safety Glasses, YAG/CO2 RK-86438-14 Mfr # LOTG-YAG/CO2
COMPARE
YAG, CO2 Green
$313.83CAD / Each
Honeywell LOTG-YAG/KTP Laser Safety Glasses, YAG/KTP RK-86438-15 Mfr # LOTG-YAG/KTP
COMPARE
YAG, harmonics Light Brown
$345.51CAD / Each
Honeywell 31-40135 Laser Safety Glasses, HeNe/Red Diode RK-86438-28 Mfr # 31-40135
COMPARE
Helium Neon (HeNe), red diodes Blue
$360.98CAD / Each