This product is now discontinued, View Related Products
Honeywell Laser Eyewear, LGF Series
Excellent visible light transmission minimizes distortion
These lightweight, impact-resistant polymer lenses are suitable for low-powered lasers or indirect laser exposure. Wavelength, optical density, and lot number are inscribed across lens for easy identification. All eyewear comes with protective case, cleaning cloth, and safety hanging cord.
LGF Series
LGF eyewear features polymer filter that protect against femtosecond (fs) and picosecond (ps) lasers. These polymer filters maintain their optical density for 10 seconds according to independent third-party testing and international standards. Eyewear features one-piece, impact-resistant polymer lens with integral molded side shields and offers a large protective area with unrestricted wide-angle viewing. Contoured frame conforms to the forehead providing excellent orbital coverage. Fits over most prescription frames.
Compliance: ANSI Z136.1-1986, Z87.1-2003 (glasses unsuitable for direct laser radiation)
$294.68 - $515.69CAD / Pair
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Laser Type Lens Color
Availability Pricing
Honeywell 31-30137 LGF Laser Eyewear, YAG/CO2 RK-86438-23 Mfr # 31-30137
COMPARE
YAG, CO2 Green
$294.68CAD / Pair
Honeywell 31-3982 Laser Safety Glasses, YAG/Diode RK-86438-24 Mfr # 31-3982
COMPARE
YAG, diode Green
$412.55CAD / Each
Honeywell 31-30102 LGF Laser Eyewear, YAG/Harmonics RK-86438-25 Mfr # 31-30102
COMPARE
YAG, harmonics Brown
$515.69CAD / Each
Honeywell 31-30135 Laser Eyewear, Helium Neon/Red Diodes RK-86438-29 Mfr # 31-30135
COMPARE
Helium Neon (HeNe), red diodes Blue
$402.68CAD / Each