This product is now discontinued, View Related Products
Hip Boots and Chest Waders
Stay warm and dry out in the field
Durable PVC exterior resists oil, grease, gasoline, saltwater, and ozone but maintains flexibility even in cold weather. Lightweight, nonabsorbent polyester lining allows easy cleaning. Hip boots and chest waders feature steel toes for impact protection, and a cleated outsole for excellent traction.
Wear over-the-sock.
Made in the USA.
Steel toe complies with ASTM F2413-11
Hip boots are 28" length to provide protection above the knee. Adjustable nylon strap keeps boots in place.
Chest waders are 54" long with top dawstring closure for a snug fit at the chest. Nylon, H-back suspenders with reinforced buttons keep waders in place. Built-in belt loop with adjustable belt for added security.
$126.71 - $213.64CAD / Pair
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Height (in) Men's Size Womens Size
Availability Pricing
Dunlop 86049 SIZE 9 Hip Boots, 28 inch Height, US Mens 9 | Womens 11 Shoe; 1 Pair RK-86420-09 Mfr # 86049 SIZE 9
COMPARE
28 9 11
$126.71CAD / Pair
Dunlop 86049 SIZE 10 Hip Boots, 28 inch Height, US Mens 10 | Womens 12 Shoe; 1 Pair RK-86420-10 Mfr # 86049 SIZE 10
COMPARE
28 10 12
$128.18CAD / Pair of 1
Dunlop 86049 SIZE 11 Hip Boots, 28 inch Height, US Mens 11 | Womens 13 Shoe; 1 Pair RK-86420-11 Mfr # 86049 SIZE 11
COMPARE
28 11 13
$126.71CAD / Pair
Dunlop 86049 SIZE 12 Hip Boots, 28 inch Height, US Mens 12 | Womens 14 Shoe; 1 Pair RK-86420-12 Mfr # 86049 SIZE 12
COMPARE
28 12 14
$128.18CAD / Pair
Dunlop 86049 SIZE 13 Hip Boots, 28 inch Height, US Mens 13 | Womens 15 Shoe; 1 Pair RK-86420-13 Mfr # 86049 SIZE 13
COMPARE
28 13 15
$126.71CAD / Pair
Dunlop 86067 SIZE 9 Chest Waders; 54 inch Height, US Mens 9 | Womens 11 Shoe; 1/Pk RK-86421-09 Mfr # 86067 SIZE 9
COMPARE
54 9 11
$210.69CAD / Pair
Dunlop 86067 SIZE 10 Chest Waders; 54 inch Height, US Mens 10 | Womens 12 Shoe; 1/Pk RK-86421-10 Mfr # 86067 SIZE 10
COMPARE
54 10 12
$213.64CAD / Pair of 1
Dunlop 86067 SIZE 11 Chest Waders; 54 inch Height, US Mens 11 | Womens 13 Shoe; 1/Pk RK-86421-11 Mfr # 86067 SIZE 11
COMPARE
54 11 13
$210.69CAD / Pair of 1
Dunlop 86067 SIZE 12 Chest Waders; 54 inch Height, US Mens 12 | Womens 14 Shoe; 1/Pk RK-86421-12 Mfr # 86067 SIZE 12
COMPARE
54 12 14
$213.64CAD / Pair of 1
Dunlop 86067 SIZE 13 Chest Waders; 54 inch Height, US Mens 13 | Womens 15 Shoe; 1/Pk RK-86421-13 Mfr # 86067 SIZE 13
COMPARE
54 13 15
$213.64CAD / Pair
Showing 1- 10 of 10