Cole-Parmer® Essentials High-Purity Solid Stoppers, Silicone
Stop particle buildup
  • Constructed of high-purity silicone to resist particle build-up
  • Autoclavable
Translucent silicone stoppers are especially suited for procedures demanding high purity, such as tissue culture work and food science applications. Food-grade quality stoppers are nontoxic, chemically inert, and resist particle buildup. Withstand repeated sterilization, as well as temperatures from –76 to 392°F (–60 to 200°C). Peak at 428°F (220°C). European size (DIN no.) is noted on stopper. FDA compliant. Autoclavable.
$24.75 - $324.88CAD / Pkg of 10
25 variations of this product are available.
See All Product Options Below
25 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
European Size Number Height (mm) Top Outside Diameter (OD) (mm)
Availability Pricing
COMPARE
6D 15 6.5
$25.64CAD / Pkg of 10
COMPARE
8D 20 8
$24.75CAD / Pkg of 10
COMPARE
10D 20 9
$27.85CAD / Pkg of 10
COMPARE
13D 20 12
$32.56CAD / Pkg of 10
COMPARE
15D 20 14.5
$33.30CAD / Pkg of 10
COMPARE
16D 20 16.5
$36.39CAD / Pkg of 10
COMPARE
18D 20 18
$41.84CAD / Pkg of 10
COMPARE
21D 25 22
$48.03CAD / Pkg of 10
COMPARE
24D 30 24
$60.41CAD / Pkg of 10
COMPARE
27D 30 27
$106.08CAD / Pkg of 10
COMPARE
29D 30 29
$117.58CAD / Pkg of 10
COMPARE
31D 30 32
$115.37CAD / Pkg of 10
COMPARE
34D 30 35
$81.33CAD / Pkg of 5
COMPARE
38D 35 38
$87.52CAD / Pkg of 5
COMPARE
41D 35 41
$123.76CAD / Pkg of 5
COMPARE
45D 40 44
$133.93CAD / Pkg of 5
COMPARE
49D 40 49
$69.69CAD / Pkg of 2
COMPARE
54D 40 55
$85.16CAD / Pkg of 2
COMPARE
60D 45 59.5
$115.37CAD / Pkg of 2
COMPARE
65D 45 65
$82.07CAD / Pkg of 1
Showing 1 - 20 of 25