This product is now discontinued, View Related Products
High Output Fiber Optic Illuminator System
Delivers 4X the light output of quartz halogen illuminators
  • 0 to 100% intensity control with manual iris
  • Reduce eye strain with greater illumination
These illuminators use a metal halide bulb to provide brilliant white light with true color representation and balance. The increased light intensity offers greater visibility and detail to your inspection and microscope applications. Adjustment of the manual iris controls light intensity.
Select single or dual self-supporting light pipes and the illuminator adapter below, to complete your system. These light pipes don't require stands or additional support. Flexible goosenecks are constructed of glass fibers sheathed in a steel monocoil. Available light filters or filter kit allows sample inspection for any need (sold separately).
$2,891.53 - $3,063.91CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Dolan Jenner 660001050100 Metal Halide Fiber Optic Illuminator, 115 VAC, 50/60 Hz RK-41724-00 Mfr # 660001050102
COMPARE
115
$2,891.53CAD / Each
Dolan Jenner 660001051100 Metal Halide Fiber Optic Illuminator, 230 VAC, 50/60 Hz RK-41724-05 Mfr # 660001051102
COMPARE
230
$3,063.91CAD / Each