Analytik Jena High-Intensity, Long-Wave UV Lamps
Detach lamp head from transformer base for portability
  • Vented housing protects against burns
These high-intensity lamps produce 365 nm UV light. Lamp head rotates 360°> in its transformer base. A filter minimizes white light interference allowing you to easily detect weak fluorescence.
$2,342.69 - $2,578.43CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Bulb / Tube Life Wavelength Description
Availability Pricing
Analytik Jena High-intensity, long-wave UV lamp; 8-ft lead, 220 VAC/50 Hz RK-97600-05 Mfr # B100AP/220V 95-0127-02
COMPARE
2000 hours 365 nm High-intensity, long-wave UV lamp; 8-ft lead, 220 VAC/50 Hz
$2,342.69CAD / Each
Analytik Jena High-intensity, long-wave UV lamp; 20-ft lead, 220 VAC/50 Hz RK-97600-15 Mfr # 95-0127-07
COMPARE
2000 hours 365 nm High-intensity, long-wave UV lamp; 20-ft lead, 220 VAC/50 Hz
$2,578.43CAD / Each