Hemco UniFlow LE AirStream Ducted Fume Hoods
Excellent low flow constant volume for energy savings and user protection
  • Maintains uniform airflow with aerodynamic face opening with air foil
  • Removable access panel allows easy maintenance of ducting connections, plumbing, and electrical services
  • 28-in. vertical sash permits convenient loading and unloading of the hood chamber
  • Meets UL 1805 classification
These fume hoods feature a nonmetallic, no-rust unitized superstructure for superior chemical resistance and increased durability. The interior chamber has all coved corners for ease of cleaning. The 3/16” clear tempered safety glass sash with frame and track fabricated of chemical-resistant PVC provide superior durability and long life. Vector air slots on back panel maintain uniform airflow through the baffle system to the bell-shaped exhaust collar outlet.
Uniflow LE AireStream Fume Hoods feature a 28” vertical sash and aerodynamic sash lift. This series also features a full 32” viewing height for working with tall equipment. These full-duty fume hoods come in 36”, 48”, 60”, 72” and 96” widths. Models are available with configurations featuring a vapor-proof light or explosion-proof light.
$12,984.99 - $21,823.85CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hood Width (in) Power (VAC) Description
Availability Pricing
HEMCO UniFlow LE AireStream Fume Hood w/ Vapor Proof Light, 36" W; 115 VAC RK-33706-52 Mfr # 35361
COMPARE
36 115 UniFlow LE AireStream Fume Hood w/ Vapor Proof Light, 36\" W; 115 VAC
$12,984.99CAD / Each
HEMCO UniFlow LE AireStream Fume Hood w/ Vapor Proof Light, 48" W; 115 VAC RK-33706-53 Mfr # 35461
COMPARE
48 115 UniFlow LE AireStream Fume Hood w/ Vapor Proof Light, 48\" W; 115 VAC
$18,191.95CAD / Each
HEMCO UniFlow LE AireStream Fume Hood w/ Vapor Proof Light, 72" W; 115 VAC RK-33706-55 Mfr # 35661
COMPARE
72 115 UniFlow LE AireStream Fume Hood w/ Vapor Proof Light, 72\" W; 115 VAC
$21,823.85CAD / Each
HEMCO UniFlow LE AireStream Fume Hood w/ Vapor Proof Light, 72" W; 230 VAC RK-33706-65 Mfr # 35664
COMPARE
72 230 UniFlow LE AireStream Fume Hood w/ Vapor Proof Light, 72\" W; 230 VAC
$21,823.85CAD / Each