Hemco UniFlow CE AirStream Ducted Fume Hoods
Excellent low flow constant volume for energy savings and user protection
  • Maintains uniform airflow with aerodynamic face opening with air foil
  • Removable access panel allows easy maintenance of ducting connections, plumbing, and electrical services
  • 23-in. vertical sash permits convenient loading and unloading of the hood chamber
  • Meets UL 1805 classification
These fume hoods feature a nonmetallic, no-rust unitized superstructure for superior chemical resistance and increased durability. The interior chamber has all coved corners for ease of cleaning. The 3/16” clear tempered safety glass sash with frame and track fabricated of chemical-resistant PVC provide superior durability and long life. Vector air slots on back panel maintain uniform airflow through the baffle system to the bell-shaped exhaust collar outlet.
Uniflow CE AireStream Fume Hoods feature a 23” vertical sash and aerodynamic sash lift. This series also features a full 27” viewing height for unobstructed view of the chamber. These full-duty fume hoods come in 30”, 36”, 48” and 72” widths. Models are available with configurations featuring a vapor-proof light or explosion-proof light as well as with or without a blower.
$12,403.00 - $21,722.19CAD / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Hood Width (in) Power (VAC) Description
Availability Pricing
HEMCO UniFlow CE AireStream Fume Hood w/ Vapor Proof Light,  30" W; 115 VAC RK-33706-28 Mfr # 13041
COMPARE
30 115 UniFlow CE AireStream Fume Hood w/ Vapor Proof Light, 30\" W; 115 VAC
$13,419.64CAD / Each
HEMCO UniFlow CE AireStream Fume Hood w/ Vapor Proof Light & Built-In Blower,  30" W; 115 VAC RK-33706-29 Mfr # 13042
COMPARE
30 115 UniFlow CE AireStream Fume Hood w/ Vapor Proof Light & Built-In Blower, 30\" W; 115 VAC
$17,511.24CAD / Each
HEMCO UniFlow CE AireStream Fume Hood w/ Vapor Proof Light,  36" W; 115 VAC RK-33706-34 Mfr # 13641
COMPARE
36 115 UniFlow CE AireStream Fume Hood w/ Vapor Proof Light, 36\" W; 115 VAC
$12,403.00CAD / Each
HEMCO UniFlow CE AireStream Fume Hood w/ Vapor Proof Light,  48" W; 115 VAC RK-33706-40 Mfr # 14841
COMPARE
48 115 UniFlow CE AireStream Fume Hood w/ Vapor Proof Light, 48\" W; 115 VAC
$14,076.77CAD / Each
HEMCO UniFlow CE AireStream Fume Hood w/ Vapor Proof Light & Built-In Blower,  48" W; 115 VAC RK-33706-41 Mfr # 14842
COMPARE
48 115 UniFlow CE AireStream Fume Hood w/ Vapor Proof Light & Built-In Blower, 48\" W; 115 VAC
$19,563.67CAD / Each
HEMCO UniFlow CE AireStream Fume Hood w/ Explosion Proof Light, 48" W; 115/230 VAC RK-33706-42 Mfr # 14843
COMPARE
48 115 / 230 UniFlow CE AireStream Fume Hood w/ Explosion Proof Light, 48\" W; 115/230 VAC
$14,076.77CAD / Each
HEMCO UniFlow CE AireStream Fume Hood w/ Vapor Proof Light,  72" W; 115 VAC RK-33706-46 Mfr # 17241
COMPARE
72 115 UniFlow CE AireStream Fume Hood w/ Vapor Proof Light, 72\" W; 115 VAC
$18,379.07CAD / Each
HEMCO UniFlow CE AireStream Fume Hood w/ Vapor Proof Light & Built-In Blower,  72" W; 115 VAC RK-33706-47 Mfr # 17242
COMPARE
72 115 UniFlow CE AireStream Fume Hood w/ Vapor Proof Light & Built-In Blower, 72\" W; 115 VAC
$21,722.19CAD / Each
HEMCO UniFlow CE AireStream Fume Hood w/ Explosion Proof Light, 72" W; 115/230 VAC RK-33706-48 Mfr # 17243
COMPARE
72 115 / 230 UniFlow CE AireStream Fume Hood w/ Explosion Proof Light, 72\" W; 115/230 VAC
$18,379.07CAD / Each
Showing 1- 9 of 9