This product is now discontinued, View Related Products
Hart Compact Calibration Baths
Model HR6330 bath delivers all the high temperature you need up to 300C. With stability and uniformity at 300C better than ± 0.015C and ± 0.020C respectively, calibrations can easily be performed at this high temperature with total uncertainty better than ± 0.05C. At lower temperatures, stability and uniformity are even better. The HR6330 is only 12 inches wide and less than 19 inches tall, so it fits easily onto a benchtop without consuming precious space.
Model HR7320 and HR7340 baths cover your needs for low temperature calibrations. The HR7320 covers a range from -20C to 150C and the HR7340 reaches even colder temperatures to -40C. Below 0C, these baths maintain an impressive stability of ± 0.005C with uniformities also better than ± 0.005C. No utility bath performs as well below 0C or at critical room and body temperatures - or even at important higher temperatures such as 100C and 122C.
For ultra cold temperatures, the HR7380 reaches -80C quickly and maintains a two sigma stability of ± 0.006C when it gets there. The HR7380 is a true metrology bath, not a chiller or circulator. With uniformity to ± 0.008C, comparison calibration of temperature devices can be performed with high precision.
Each bath includes an RS232 serial interface and HR9930 interface-it software for controlling your bath from a PC. With a Hart Scientific thermometer readout, such as a Black Stack, and the Met Temp II software, automated calibrations can run unattended.
$14,389.13 - $30,432.87CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Power (VAC)
Availability Pricing
Fluke Calibration 6330 Compact Temperature Calibration Bath, 35C to 300C RK-16101-45 Mfr # 6330
COMPARE
572 95 115
$17,573.12CAD / Each
Fluke Calibration 7320 Fluke (Hart) Compact Temperature Calibration Bath, -20 to 150C RK-16101-46 Mfr # 7320
COMPARE
$14,389.13CAD / Each
Fluke Calibration 7340 Compact Temperature Calibration Bath, -40 to 150C RK-16101-47 Mfr # 7340
COMPARE
302 -40 115
$24,090.53CAD / Each
Fluke Calibration 7380 Compact Temperature Calibration Bath, -80 to 100C RK-16101-48 Mfr # 7380
COMPARE
212 -112 115
$30,432.87CAD / Each