This product is now discontinued, View Related Products
Hand-Motion Shaker
Ideal for DNA extractions, pretreatment of enzyme reactions, treatment of reagents, and gel treatment for electrophoresis
Perform continuous or timed treatments using built-in timer. Adjust shaker platform for a more vertical or more horizontal shaking angle, giving you ability to customize for your specific applications. Two adhesive rubber pads are located at the base. Shaker requires one of four platforms sold separately.
$1,782.80 - $1,803.43CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (VAC)
Availability Pricing
Hand-Motion Shaker, 0 to 45° angle, 60 to 180 rpm, 115 VAC RK-04636-60 Mfr # SHK-COCK, 115V
COMPARE
115
$1,803.43CAD / Each
Hand-Motion Shaker, 0 to 45° angle, 60 to 180 rpm, 230 VAC RK-04636-65 Mfr # SHK-COCK, 230V
COMPARE
230
$1,782.80CAD / Each