This product is now discontinued, View Related Products
Haier Upright Refrigerators
High efficiency refrigeration system for optimal performance
  • Forced-air cooling with internal fan for maximum temperature uniformity and stability
  • Cooling fan is permanently lubricated for long life
  • Automatic defrost maintains optimum cooling capacity
  • Multiple alarm system for safe and reliable operation
  • Glass door with electric heat reduces condensation
Designed to meet the demanding requirements of everyday use, these refrigerators incorporate the latest in cabinet, refrigeration, and temperature control features to provide energy efficient, safe, convenient, and reliable storage of temperature sensitive items in a variety of applications. A microprocessor controller has push button control and a digital temperature display. A complete temperature alarm system features an audible buzzer and visible flashing light that will alert for failure due to high or low temperature, sensor error, door ajar, low battery, or power failure. Each refrigerator has a bright interior light, external port, safety lock and keys, and leveling legs and casters.
15 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
Haier HYC-290(220V/50Hz) 10.2 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 220 VAC, 50 Hz RK-44260-37 Mfr # HYC-290(220V/50Hz)
COMPARE
50 220 10.2
$6,247.17CAD / Each
Haier HYC-290(220V/60Hz) 10.2 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 220 VAC, 60 Hz RK-44260-38 Mfr # HYC-290(220V/60Hz)
COMPARE
60 220 10.2
$6,247.17CAD / Each
Haier HYC-290(115V/60Hz) 10.2 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 115 VAC, 60 Hz RK-44260-39 Mfr # HYC-290(115V/60Hz)
COMPARE
60 115 10.2
$6,247.17CAD / Each
Haier HYC-390(220V/50Hz) 13.8 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 220 VAC, 50 Hz RK-44260-41 Mfr # HYC-390(220V/50Hz)
COMPARE
50 220 13.8
$7,447.99CAD / Each
Haier HYC-390(220V/60Hz) 13.8 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 220 VAC, 60 Hz RK-44260-42 Mfr # HYC-390(220V/60Hz)
COMPARE
60 220 13.8
$6,947.03CAD / Each
Haier HYC-390(115V/60Hz) 13.8 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 115 VAC, 60 Hz RK-44260-43 Mfr # HYC-390(115V/60Hz)
COMPARE
60 115 13.8
$7,447.99CAD / Each
Haier HYC-610(220V/50Hz) 21.6 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 220 VAC, 50 Hz RK-44260-44 Mfr # HYC-610(220V/50Hz)
COMPARE
50 220 21.6
$11,941.83CAD / Each
Haier HYC-610(220V/60Hz) 21.6 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 220 VAC, 60 Hz RK-44260-45 Mfr # HYC-610(220V/60Hz)
COMPARE
60 220 21.6
$11,941.83CAD / Each
Haier HYC-610(115V/60Hz) 21.6 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 115 VAC, 60 Hz RK-44260-46 Mfr # HYC-610(115V/60Hz)
COMPARE
60 115 21.6
$11,941.83CAD / Each
Haier HYC-940(220V/50Hz) 31.4 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 220 VAC, 50 Hz RK-44260-47 Mfr # HYC-940(220V/50Hz)
COMPARE
50 220 31.4
$14,291.88CAD / Each
Haier HYC-940(220V/60Hz) 31.4 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 208-230 VAC, 60 Hz RK-44260-48 Mfr # HYC-940(220V/60Hz)
COMPARE
60 208 / 230 31.4
$14,291.88CAD / Each
Haier HYC-940(115V/60Hz) 31.4 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 115 VAC, 60 Hz RK-44260-49 Mfr # HYC-940(115V/60Hz)
COMPARE
60 115 31.4
$15,875.78CAD / Each
Haier HYC-1378(220V/50Hz) 48.7 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 220 VAC, 50 Hz RK-44260-51 Mfr # HYC-1378(220V/50Hz)
COMPARE
50 220 48.7
$14,453.96CAD / Each
Haier HYC-1378(220V/60Hz) 48.7 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 208-230 VAC, 60 Hz RK-44260-52 Mfr # HYC-1378(220V/60Hz)
COMPARE
60 208 / 230 48.7
$13,488.89CAD / Each
Haier HYC-1378(115V/60Hz) 48.7 Cu Ft 2-8℃ Upright Pharmacy Refrigerator, 115 VAC, 60 Hz RK-44260-53 Mfr # HYC-1378(115V/60Hz)
COMPARE
60 115 48.7
$14,453.96CAD / Each
Showing 1- 15 of 15