Haier Undercounter Refrigerators
Safe cold storage in a convenient undercounter size
  • Forced-air cooling with internal fan for maximum temperature uniformity and stability
  • Automatic defrost maintains optimum cooling capacity
  • Multiple alarm system for safe and reliable operation
Designed to meet the demanding requirements of everyday use, these compact refrigerators incorporate the latest in cabinet, refrigeration, and temperature control features to provide energy efficient, safe, convenient, and reliable storage of temperature sensitive items in a variety of applications. A microprocessor controller has push button control and a digital temperature display. A complete temperature alarm system features an audible buzzer and visible flashing light that will alert for failure due to high or low temperature, sensor error, door ajar, or power failure. Available with either a glass or solid door each refrigerator has a bright interior LED, external port, and a safety lock to prevent unauthorized access.
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Doors Power (VAC) Refrigerator Capacity (cu ft)
Availability Pricing
Haier HYC-118A(115V/60Hz) 4.2 Cu Ft 2-8℃ Glass Door Undercounter Pharmacy Refrigerator, 115 VAC, 60 Hz RK-44260-36 Mfr # HYC-118A(115V/60Hz)
COMPARE
1 Glass 115 4.2
$3,676.11CAD / Each