This product is now discontinued, View Related Products
Hach Pocket Colorimeter II Kits
Portable single-parameter photometers go anywhere!
  • Simple to use with fast, accurate results
  • Logs last ten data points with measurement time
  • Rugged, IP67-rated waterproof housing ideal for field testing
The Hach Pocket II single-parameter colorimeters have an easy-to-read backlit display and a four-button keypad that allow for easy operation. Many of the factory programmed parameters are based on EPA-approved methods. No manual calibration is required, simply zero the instrument with a blank, insert the reacted sample, and read the result.
$692.49 - $1,897.73CAD / Each
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
Compareitem specifications
Detector Type Test Parameters Wavelength(s) (nm)
Availability Pricing
Hach 5870025 Pocket Colorimeter Ii, Aluminum RK-99574-30 Mfr # 5870025
COMPARE
Silicon Photodiode Aluminum Fixed
$772.06CAD / Each
Hach 5870040 Pocket Colorimeter Ii, Ammonia 2 1/2" x 6 3/16" x 1 3/8" RK-99574-32 Mfr # 5870040
COMPARE
Silicon Photodiode Ammonia Fixed
$772.06CAD / Each
Hach 5870026 Pocket Colorimeter Ii, Ammonia, Mono RK-99574-34 Mfr # 5870026
COMPARE
Silicon Photodiode Ammonia and Monochloramine Fixed
$745.54CAD / Each
Hach 5870001 Pocket Colorimeter Ii, Bromine 2 1/2" x 6 3/16" x 1 3/8" RK-99574-36 Mfr # 5870001
COMPARE
Silicon Photodiode Bromine Fixed
$745.54CAD / Each
Hach 5870051 Pocket Colorimeter Ii, Chlorine Dioxide RK-99574-40 Mfr # 5870051
COMPARE
Silicon Photodiode Chlorine dioxide Fixed
$772.06CAD / Each
Hach 5870023 Pocket Colorimeter Ii, Chlorine, Free RK-99574-42 Mfr # 5870023
COMPARE
Silicon Photodiode Chlorine; free Fixed
$760.27CAD / Each
Hach 5870024 Pocket Colorimeter II, Chlorine, Total RK-99574-44 Mfr # 5870024
COMPARE
Silicon Photodiode Chlorine; total Fixed
$745.54CAD / Each
Hach 5870000 Pocket Colorimeter Ii, Chlorine, Free and Total RK-99574-46 Mfr # 5870000
COMPARE
Silicon Photodiode Chlorine; free and total Fixed
$692.49CAD / Each
Hach 5870017 Pocket Colorimeter Iii, Chromium, Hexavalent RK-99574-56 Mfr # 5870017
COMPARE
Silicon Photodiode Chromium hexavalent Fixed
$745.54CAD / Each
Hach 5870019 Pocket Colorimeter Ii, Copper (LCD) RK-99574-58 Mfr # 5870019
COMPARE
Silicon Photodiode Copper Fixed
$772.06CAD / Each
Hach 5870005 Pocket Colorimeter Ii, Fluoride RK-99574-62 Mfr # 5870005
COMPARE
Silicon Photodiode Fluoride Fixed
$794.16CAD / Each
Hach 5870022 Pocket Colorimeter IiIron 2 1/2" x 6 3/16" x 1 3/8" RK-99574-66 Mfr # 5870022
COMPARE
Silicon Photodiode Iron Fixed
$745.54CAD / Each
Hach 5870016 Pocket Colorimeter IiIron, TPTZ RK-99574-68 Mfr # 5870016
COMPARE
Silicon Photodiode Iron and TPTZ Fixed
$745.54CAD / Each
Hach 5870021 Pocket Colorimeter Ii, Lead 2 1/2" x 6 3/16" x 1 3/8" RK-99574-70 Mfr # 5870021
COMPARE
Silicon Photodiode Lead Fixed
$972.44CAD / Each
Hach 5870015 Pocket Colorimeter Ii, Manganese, HR RK-99574-72 Mfr # 5870015
COMPARE
Silicon Photodiode Manganese; high-range Fixed
$745.54CAD / Each
Hach 5870018 Pocket Colorimeter Ii, Manganese LR RK-99574-74 Mfr # 5870018
COMPARE
Silicon Photodiode Manganese; low-range Fixed
$745.54CAD / Each
Hach 5870010 Pocket Colorimeter Ii, Molybdate RK-99574-76 Mfr # 5870010
COMPARE
Silicon Photodiode Molybdate Fixed
$745.54CAD / Each
Hach 5870020 Pocket Colorimeter Ii, Nickel and Cobalt RK-99574-78 Mfr # 5870020
COMPARE
Silicon Photodiode Nickel and Cobalt Fixed
$745.54CAD / Each
Hach 5870002 Pocket Colorimeter Ii, Nitrate 2 1/2" x 6 3/16" x 1 3/8" RK-99574-80 Mfr # 5870002
COMPARE
Silicon Photodiode Nitrate Fixed
$772.06CAD / Each
Hach 5870004 Pocket Colorimeter Ii, Ozone 2 1/2" x 6 3/16" x 1 3/8" RK-99574-82 Mfr # 5870004
COMPARE
Silicon Photodiode Ozone Fixed
$772.06CAD / Each
Showing 1 - 20 of 26