This product is now discontinued, View Related Products
Hach HQ30d Field LDO, pH/LDO and Conductivity/LDO Meter Kits
Convenience and accuracy of luminescent dissolved oxygen technology
  • IntellCAL™ smart probes have “plug-and-play: technology for maximum flexibility
  • Measure dissolved oxygen or BOD using maintenance-free LDO technology
  • Option to select from 13 languages in setup mode
  • Store up to 500 data sets with GLP-compliance
  • Rugged, IP67-rated waterproof housing
Versatile HQd meters offer a large backlit display with an intuitive user-interface. The meter prompts you through calibration, setup, and measurements; features single input with multiparameter operation for LDO, pH, conductivity, and temperature.
Meter can automatically or manually log up to 500 measurements with probe serial number, current calibration data, user ID, sample ID, time, and date. Use the USB port to download data to a PC or flash drive.
Precalibrated Hach IntelliCAL smart probes store their own calibration data and identification. The meter recognizes each probe, so you can easily switch probes, and the meter is automatically setup to run the next parameter without recalibrating. Kits include rugged probe for field applications; all connections between the meter and probe are secure and waterproof. Order optional laboratory probe separately below.
$3,237.04 - $4,831.25CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Conductivity Max Dissolved Oxygen Max pH (pH)
Availability Pricing
Hach 8505100 HQ30d Digital LDO Portable Field Meter Kit RK-59004-40 Mfr # 8505100
COMPARE
$3,237.04CAD / Each
Hach 8505400 HQ30d Digital pH/LDO Portable Field Meter Kit RK-59004-42 Mfr # 8505400
COMPARE
14
$4,831.25CAD / Each
Hach 8507500 HQ30d Digital Conductivity/LDO Portable Field Meter Kit RK-59004-46 Mfr # 8507500
COMPARE
$4,831.25CAD / Each