This product is now discontinued, View Related Products
H-B Instrument Standard Enviro-Safe Nonhazardous Pocket Glass Thermometers, Fahrenheit-scale
Rugged case with an eyelet at the top for easy suspension
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
The case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation (except closed case styles). Available in three case styles; closed metal, open metal, closed plastic, or open plastic. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant and have a statement of accuracy.
$26.52 - $45.68CAD / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Overall Length (mm)
Availability Pricing
H-B Instrument B60570-1500 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 0 to 220F, Closed Metal Case, Environmentally Friendly RK-08007-03 Mfr # 605701500
COMPARE
220 0 160
$29.47CAD / Each
H-B Instrument B60570-1600 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 20 to 120F, Closed Metal Case, Environmentally Friendly RK-08007-04 Mfr # 605701600
COMPARE
120 20 160
$29.47CAD / Each
H-B Instrument B60570-1100 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 0 to 220F, Window Metal Case, Environmentally Friendly RK-08008-82 Mfr # 605701100
COMPARE
220 0 160
$32.41CAD / Each
H-B Instrument B60570-1200 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 20 to 120F, Window Metal Case, Environmentally Friendly RK-08008-83 Mfr # 605701200
COMPARE
120 20 160
$32.41CAD / Each
H-B Instrument B60570-0700 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 0 to 220F, Closed Plastic Case, Environmentally Friendly RK-08008-86 Mfr # 605700700
COMPARE
220 0 160
$26.52CAD / Each
H-B Instrument B60570-0800 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 20 to 120F, Closed Plastic Case, Environmentally Friendly RK-08008-87 Mfr # 605700800
COMPARE
120 20 160
$26.52CAD / Each
H-B Instrument B60570-0300 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 0 to 220F, Window Plastic Case, Environmentally Friendly RK-08008-90 Mfr # 605700300
COMPARE
220 0 160
$26.52CAD / Each
H-B Instrument B60570-0400 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 20 to 120F, Window Plastic Case, Environmentally Friendly RK-08008-91 Mfr # 605700400
COMPARE
120 20 160
$26.52CAD / Each
H-B Instrument B60570-2000 Enviro-Safe Liquid-In-Glass Pocket Thermometer; 0 to 220F, Aluminum Duplex Case, Environmentally Friendly RK-08008-94 Mfr # 605702000
COMPARE
220 0 160
$45.68CAD / Each
H-B Instrument Enviro-Safe Liquid-In-Glass Pocket Laboratory Thermometer; 20 to 120°F, Closed Metal Case, Environmentally Friendly RK-08008-95 Mfr # 605702100
COMPARE
120 20 160
$45.68CAD / Each
H-B Instrument Enviro-Safe Pocket Thermometer, Nickel Plated BRASS Case, 0/220F, 160MM LENGTH RK-08008-98 Mfr # B60570-2400
COMPARE
219 0 160
$41.25CAD / Each
H-B Instrument Enviro-Safe Pocket Thermometer, Nickel Plated BRASS Case, 20/120F, 160MM LENGTH RK-08008-99 Mfr # B60570-2500
COMPARE
120 19 160
$39.41CAD / Each
Showing 1- 12 of 12