This product is now discontinued, View Related Products
H-B Instrument Enviro-Safe Nonhazardous General-Purpose Glass Thermometers, Fahrenheit-scale
The only thermometers certified as 100% non-toxic, non-hazardous, biodegradable, and “green” !
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy
These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater and many other general purpose applications where the use of mercury or other hazardous liquids are not desired. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant, anti-roll fitting, individual serial numbered and have a statement of accuracy.
$20.77 - $48.62CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Overall Length (mm)
Availability Pricing
H-B Instrument B60501-0200 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 20 to 230F, 50mm Immersion, Environmentally Friendly RK-08008-61 Mfr # B60501-0200
COMPARE
230 20 200
$21.07CAD / Each
H-B Instrument B60504-0200 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 20 to 230F, Total Immersion, Environmentally Friendly RK-08008-62 Mfr # B60504-0200
COMPARE
230 20 200
$20.77CAD / Each
H-B Instrument B60501-0300 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 20 to 300F, 50mm Immersion, Environmentally Friendly RK-08008-63 Mfr # B60501-0300
COMPARE
300 20 200
$21.07CAD / Each
H-B Instrument B60504-0400 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 20 to 300F, Total Immersion, Environmentally Friendly RK-08008-64 Mfr # B60504-0400
COMPARE
300 20 200
$21.07CAD / Each
H-B Instrument B60502-0400 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 0 to 230F, 76mm Immersion, Environmentally Friendly RK-08008-69 Mfr # B60502-0400
COMPARE
230 0 300
$23.87CAD / Each
H-B Instrument B60506-0400 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 0 to 230F, Total Immersion, Environmentally Friendly RK-08008-70 Mfr # B60506-0400
COMPARE
230 0 300
$22.98CAD / Each
H-B Instrument B60502-0500 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 0 to 300F, 76mm Immersion, Environmentally Friendly RK-08008-71 Mfr # B60502-0500
COMPARE
300 0 300
$34.99CAD / Each
H-B Instrument B60506-0500 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 0 to 300F, Total Immersion, Environmentally Friendly RK-08008-72 Mfr # B60506-0500
COMPARE
300 0 300
$25.49CAD / Each
H-B Instrument B60503-0100 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 20 to 500F, 76mm Immersion, Environmentally Friendly RK-08008-75 Mfr # B60503-0100
COMPARE
500 20 350
$38.31CAD / Each
H-B Instrument B60507-0100 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; 20 to 440F, Total Immersion, Environmentally Friendly RK-08008-76 Mfr # B60507-0100
COMPARE
440 20 350
$34.62CAD / Each
H-B Instrument B60502-0300 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -30 to 120F, 76mm Immersion, Environmentally Friendly RK-90300-50 Mfr # 605020300
COMPARE
120 -30 300
$48.62CAD / Each
Showing 1- 11 of 11