This product is now discontinued, View Related Products
H-B Instrument Enviro-Safe Nonhazardous General-Purpose Glass Thermometers, Celsius-scale
The only thermometers certified as 100% non-toxic, non-hazardous, biodegradable, and “green” !
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy
These thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater and many other general purpose applications where the use of mercury or other hazardous liquids are not desired. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant, anti-roll fitting, individual serial numbered and have a statement of accuracy.
$14.73 - $48.62CAD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
H-B Instrument B60501-0000 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -10 to 110C, 50mm Immersion, Environmentally Friendly RK-08008-57 Mfr # B60501-0000
COMPARE
110 -10 200
$21.66CAD / Each
H-B Instrument B60504-0000 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -10 to 110C, Total Immersion, Environmentally Friendly RK-08008-58 Mfr # B60504-0000
COMPARE
110 -10 200
$19.89CAD / Each
H-B Instrument B60501-0100 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -10 to 150C, 50mm Immersion, Environmentally Friendly RK-08008-59 Mfr # B60501-0100
COMPARE
150 -10 200
$19.89CAD / Each
H-B Instrument B60504-0100 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -10 to 150C, Total Immersion, Environmentally Friendly RK-08008-60 Mfr # B60504-0100
COMPARE
150 -10 200
$19.89CAD / Each
H-B Instrument B60502-0100 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -20 to 110C, 76mm Immersion, Environmentally Friendly RK-08008-65 Mfr # B60502-0100
COMPARE
110 -20 300
$26.08CAD / Each
H-B Instrument B60506-0100 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -20 to 110C, Total Immersion, Environmentally Friendly RK-08008-66 Mfr # B60506-0100
COMPARE
110 -20 300
$32.05CAD / Each
H-B Instrument B60502-0200 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -20 to 150C, 76mm Immersion, Environmentally Friendly RK-08008-67 Mfr # B60502-0200
COMPARE
150 -20 300
$27.41CAD / Each
H-B Instrument B60506-0200 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -20 to 150C, Total Immersion, Environmentally Friendly RK-08008-68 Mfr # B60506-0200
COMPARE
150 -20 300
$26.82CAD / Each
H-B Instrument B60503-0000 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -10 to 260C, 76mm Immersion, Environmentally Friendly RK-08008-73 Mfr # B60503-0000
COMPARE
260 -10 350
$37.57CAD / Each
H-B Instrument B60507-0000 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -10 to 225C, Total Immersion, Environmentally Friendly RK-08008-74 Mfr # B60507-0000
COMPARE
225 -10 350
$36.10CAD / Each
H-B Instrument B60502-0000 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -35 to 50C, 76mm Immersion, Environmentally Friendly RK-90300-49 Mfr # 605020000
COMPARE
50 -35 300
$36.83CAD / Each
H-B Instrument B60506-0000 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -35 to 50C, Total Immersion, Environmentally Friendly RK-90300-51 Mfr # 605060000
COMPARE
50 -35 300
$48.62CAD / Each
H-B Instrument B60506-0300 Enviro-Safe General Purpose Liquid-In-Glass Thermometer; -30 to 120F, Total Immersion, Environmentally Friendly RK-90300-52 Mfr # B60506-0300
COMPARE
$14.73CAD / Each
Showing 1- 13 of 13