This product is now discontinued, View Related Products
H-B Instrument Easy-Read Enviro-Safe Nonhazardous Pocket Glass Thermometers, Celsius-scale
Rugged case with an eyelet at the top for easy suspension
  • Enviro-Safe liquid within the thermometers is non-toxic, non-hazardous, and fully biodegradable
  • Replace mercury thermometers with the same accuracy without any hazards
  • Statement of accuracy included
The case protects these thermometers from accidental breakage during use and storage. The section of the case that surrounds the bulb has small openings for circulation (except closed case styles). Available in two case styles; closed metal, or open metal. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. Each thermometer features a green liquid and white back glass. These thermometers also feature recyclable packaging, RoHS compliant and have a statement of accuracy.
Easy-Read® thermometers feature a black liquid and yellow back glass - distinct contrast makes these thermometers very easy to read.
$29.84 - $34.26CAD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
H-B Instrument B60370-0100 Easy-Read Pocket Liquid-In-Glass Thermometer; -10 to 110C, Closed Metal Case, Environmentally Friendly RK-90300-37 Mfr # B60370-0100
COMPARE
110 -10 160
$33.15CAD / Each
H-B Instrument B60370-0200 Easy-Read Pocket Liquid-In-Glass Thermometer; -5 to 50C, Closed Metal Case, Environmentally Friendly RK-90300-38 Mfr # B60370-0200
COMPARE
50 -5 160
$33.89CAD / Each
H-B Instrument B60370-0400 Easy-Read Pocket Liquid-In-Glass Thermometer; -30 to 50C, Closed Metal Case, Environmentally Friendly RK-90300-40 Mfr # B60370-0400
COMPARE
50 -30 160
$29.84CAD / Each
H-B Instrument EASY-READ POCKET Thermometer, Window NICKLE-PLATED BRASS Case, -5/50C RK-90300-41 Mfr # B60370-1200
COMPARE
50 -5 160
$34.26CAD / Each
H-B Instrument B60370-0600 Easy-Read Pocket Liquid-In-Glass Thermometer; -10 to 110C, Window Metal Case, Environmentally Friendly RK-90300-43 Mfr # B60370-0600
COMPARE
110 -10 160
$33.15CAD / Each
H-B Instrument B60370-0700 Easy-Read Pocket Liquid-In-Glass Thermometer; -5 to 50C, Window Metal Case, Environmentally Friendly RK-90300-44 Mfr # B60370-0700
COMPARE
50 -5 160
$33.89CAD / Each
H-B Instrument B60370-0900 Easy-Read Pocket Liquid-In-Glass Thermometer; -30 to 50C, Window Metal Case, Environmentally Friendly RK-90300-46 Mfr # B60370-0900
COMPARE
50 -30 160
$29.84CAD / Each
H-B Instrument B60370-1100 Easy-Read Pocket Liquid-In-Glass Thermometer; -5 to 50C, Window Plastic Case, Environmentally Friendly RK-90300-48 Mfr # B60370-1100
COMPARE
50 -5 160
$31.31CAD / Each
Showing 1- 8 of 8