This product is now discontinued, View Related Products
Cole-Parmer API Plain Form Hydrometers
Quick and straightforward petroleum density measurements
  • Plain form simplifies readings
  • Individually serialized for easy identification
  • Lead-free ballast and glass construction nontoxic to clean up after breakage
  • Temperature standard is 60°F (15.6°C)
$69.25 - $78.09CAD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem Availability Pricing
COMPARE
$70.72CAD / Each
COMPARE
$78.09CAD / Each
COMPARE
$70.72CAD / Each
COMPARE
$70.72CAD / Each
COMPARE
$70.72CAD / Each
COMPARE
$70.72CAD / Each
COMPARE
$70.72CAD / Each
COMPARE
$69.25CAD / Each
COMPARE
$69.25CAD / Each
COMPARE
$78.09CAD / Each
COMPARE
$70.72CAD / Each
Showing 1- 11 of 11