This product is now discontinued, View Related Products
H-B Instrument Angled Storage Trays for Glass Thermometers
Designed to properly store your glass thermometers at the correct angle
  • Use for all types of liquid-in-glass thermometers
  • Angled to prevent separation of liquid column
  • UV coated
Keep thermometers handy and properly stored while keeping your space organized. Trays are made of 2 mm white polystyrene with an inclined angle to help prevent liquid separation. Each tray has numbered slots. Choose from two sizes - one fits easily in a drawer, the other is suited for table for shelf storage. Sold in sets.
$104.61 - $115.81CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
H-B Instrument B61400-5300 Angled Liquid-in-Glass Thermometer Storage Tray; 3 Slots RK-08007-96 Mfr # B61400-5300
COMPARE
Angled Liquid-in-Glass Thermometer Storage Tray; 3 Slots
$104.61CAD / Each
H-B Instrument B61400-5100 Angled Liquid-in-Glass Thermometer Storage Tray; 14 Slots RK-08007-97 Mfr # 614005100
COMPARE
Angled Liquid-in-Glass Thermometer Storage Tray; 14 Slots
$115.81CAD / Each