GW Instek PSB Series Programmable Switching DC Power Supplies
Precision power in a compact design
  • 90-degree vertical or horizontal rotating interface optimizes bench space
  • Constant power output provides multirange voltage and current operation
  • Connect up to two units in series or up to four units in parallel for increased power
  • Quickly and easily recall regularly used current/voltage settings with three sets of preset function keys
  • Overvoltage and overcurrent functions ensure device under test protection
  • Standard RS-232 and USB interfaces with optional GPIB
$3,789.56CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek PSB-2400L2 DC Power Supply 2CH, 80 V, 40 A, 400 W RK-20050-35 Mfr # PSB-2400L2
COMPARE
DC Power Supply 2CH, 80 V, 40 A, 400 W
$3,789.56CAD / Each