This product is now discontinued, View Related Products
GW Instek GDM-8255A/8251A 5-1/2-Digit Dual-Display Benchtop Multimeters
Save time and money on debugging and analysis
  • Two-wire and four-wire resistance measurements for low and high current requirements
  • User-selectable measurement speed rates maximize measurement efficiency and accuracy
  • RS-232, USB, and digital I/O provide added connectivity flexibility
  • Included driver and PC software for remote operation and powerful graphical analysis
  • Optional eighteen-channel scanner card eliminates the complexities of multipoint measurements and data processing (GDM-8255A only)
These high-resolution multimeters allow you to read even the smallest signals. Units feature DC voltage accuracy of ±0.012% with an industry-leading response time, allowing you to quickly see precise measurements. Advanced functions include simultaneous dual display measurements and a convenient remote control function for ease of use and additional flexibility. Nine measuring functions and ten advanced functions greatly enhance the effectiveness of measuring results and digital I/O, USB, and RS-232 provide options for connectivity.
$898.77 - $1,150.72CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GW Instek GDM-8251A Multimeter, Benchtop, 5.5" Digit, 120000 Counts RK-20050-47 Mfr # GDM-8251A
COMPARE
Multimeter, Benchtop, 5.5\" Digit, 120000 Counts
$898.77CAD / Each
GW Instek GDM-8255A Multimeter, Benchtop,  5.5" Digit, 199999 Counts RK-20050-48 Mfr # GDM-8255A
COMPARE
Multimeter, Benchtop, 5.5\" Digit, 199999 Counts
$1,150.72CAD / Each