This product is now discontinued, View Related Products
Groundwater Filter Capsules
Filter groundwater samples for dissolved metal analysis
Groundwater filter capsules are designed to make the process of collecting groundwater samples for analysis quick, easy, and convenient. Capsules contain no glues, adhesives, or other surfactants. All housing components are thermally fused. Capsules provide at least 720 cm2 of effective filtration area. Connections are 1/4 to 3/8" (6 to 9 mm) stepped barb.
$92.68 - $142.99CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Pore Size (micron)
Availability Pricing
ZenPure Ground water filter capsule, 0.45 um (1/4" to 3/8" stepped hosebarb) RK-49705-92 Mfr # SKLS045H2HSHS
COMPARE
0.45
$142.99CAD / Each
ZenPure Ground water filter capsule, 1.0um (1/4" to 3/8" stepped hosebarb) RK-49705-94 Mfr # SKLP010H2HSHS
COMPARE
1
$103.73CAD / Each
ZenPure Ground water filter capsule, 5.0um (1/4" to 3/8" stepped hosebarb) RK-49705-96 Mfr # SKLP050H2HS2HS
COMPARE
5
$92.68CAD / Each