Cole-Parmer® Graphite Straight Ferrules for Packed Columns
Reusable one-piece design
  • Constructed of durable graphite
Graphite Ferrules are preferred for capillary columns. Maximum temperature: 450degC. Does not stick to glass and is reusable if not over-tightened. One-piece design – no back ferrule.
$61.15CAD / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Graphite straight ferrules, 1/8\"; 10/pack
$61.15CAD / Pkg of 10
COMPARE
Graphite straight ferrules, 1/4\"; 10/pack
$61.15CAD / Pkg of 10
COMPARE
Graphite straight ferrules, 1/16\"; 10/pack
$61.15CAD / Pkg of 10