Corning® Gosselin™ Sterile Disposable Cell Spreaders, Polystyrene
Safe, efficient sample plating
  • Sterile polystyrene (PS)—no need to preclean
  • Eliminate the risk of cross-contamination
  • Available in L shape and T shape to cover all uses
  • Conveniently packaged in resealable bags
These sterile disposable cell spreaders are ideal for evenly spreading bacterial cultures while reducing the risk of cross contamination. Spreaders are color coded for easy identification.
L-shaped spreader quickly swirls the culture sample around the petri dish.
T-shaped spreader offers more precise maneuverability for detailed spreading protocols.
$1,259.75 - $1,856.47CAD / Case of 6000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Handle Type Overall Length (mm)
Availability Pricing
Corning Gosselin Cell Spreader, Sterile, Polystyrene, L-Shaped, White, 148 mm L; 6000/Cs RK-34551-46 Mfr # ETAR-05
COMPARE
White L-Shaped 148
$1,856.47CAD / Case of 6000
Corning Gosselin Cell Spreader, Sterile, Polystyrene, T-Shaped, Green, 150 mm L; 3500/Cs RK-34551-48 Mfr # ETAT-04
COMPARE
Green T-Shaped 150
$1,259.75CAD / Case of 3500