This product is now discontinued, View Related Products
Corning® Gosselin™ Blender Bags
PE Sample blender bags with optional side filter
  • Use as universal bags for any of your applications
  • Without or with a non woven filter for reduced particles
  • Each filter bag has a printed lot number
  • Smart choice for suspension sampling with pipets
  • Smart choice for homogenous suspension
  • Sterile
Use Gosselin™ blender bags with the Gosselin Adjustable Sample Blender (catalog MM-34551-08) or as universal bags. The sterile PE material sample bags are available in 80 mL and 400 mL sizes. The sample bags are available without and with a side filter to reduce particles from a primary dilution stage. The bag design improves the pipetting process after homogenization. The closure clip is sold separately with catalog number MM-34551-65.
$609.62 - $1,694.40CAD / Case of 4500
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Bag Capacity (mL) Description
Availability Pricing
Corning Gosselin SM1-01 Blender Bag without Filter, polyethylene, 80 mL; 4500/cs RK-34551-39 Mfr # SM1-01
COMPARE
80 Blender Bag without Filter, polyethylene, 80 mL; 4500/pk
$1,535.27CAD / Case of 4500
Corning Gosselin SM2-01 Blender Bag without Filter, polyethylene, 400 mL; 2500/cs RK-34551-40 Mfr # SM2-01
COMPARE
400 Blender Bag without Filter, polyethylene, 400 mL; 2500/pk
$1,117.57CAD / Case of 2500
Corning Gosselin SM3-01 Blender Bag without Filter, polyethylene, 3.5 L; 500/pk RK-34551-41 Mfr # SM3-01
COMPARE
3500 Blender Bag without Filter, polyethylene, 3.5 L; 500/pk
$609.62CAD / Case of 500
Corning Gosselin BBAG-03 Blender Bag with Lateral Filter, polyethylene, 400 mL; 1500/cs; 3 boxes x 20 bags of 25 RK-34551-42 Mfr # BBAG-03
COMPARE
400 Blender Bag with Lateral Filter, polyethylene, 400 mL, without logo; 1500/pk
$1,694.40CAD / Case of 1500
Corning Gosselin BBAG-04 Blender Bag with Lateral Filter, polyethylene, 400 mL; 1500/cs; 3 boxes x 10 bags of 50 RK-34551-43 Mfr # BBAG-04
COMPARE
400 Blender Bag with Lateral Filter, polyethylene, 400 mL, without symbol; 1500/pk
$1,626.62CAD / Case of 1500
Corning Gosselin FBAG-03 Blender Bag with Full Filter, polyethylene, 400 mL; 1500/cs; 3 boxes x 20 bags of 25 RK-34551-44 Mfr # FBAG-03
COMPARE
400 Blender Bag with Lateral Filter, polyethylene, 400 mL, without symbol; 1500/pk
$1,694.40CAD / Case of 1500
Corning Gosselin FBAG-04 Blender Bag with Full Filter, polyethylene, 400 mL; 1500/cs; 3 boxes x 10 bags of 50 RK-34551-45 Mfr # FBAG-04
COMPARE
400 Blender Bag with Full Filter, polyethylene, 400 mL, without symbol; 1500/pk
$1,626.62CAD / Case of 1500
Showing 1- 7 of 7