Glass Trays

Pyrex 3170 heavy duty sterilizing tray is designed to endure hot air or steam sterilization. Pyrex 3175 trays are ideal for drying, staining, or developing and are autoclavable; trays have handles for easy ausage. Both tray models can be used with electophoresis gels and paper chromatography. Made for oven use only; do not place over direct heat.
$154.71 - $1,915.41CAD / Case of 4

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L)
Availability Pricing
Pyrex 3175-7 Brand 3175 Glass Drying Tray; 275mm L x 175mm W x 37mm H, 4/Cs RK-34588-20 Mfr # 3175-7
COMPARE
2
$154.71CAD / Case of 4
Pyrex 3175-8 Brand 3175 Glass Drying Tray; 200mm L x 200mm W x 50mm H, 4/Cs RK-34588-22 Mfr # 3175-8
COMPARE
2
$154.71CAD / Case of 4
Pyrex 3175-10 Brand 3175 Glass Drying Tray; 349mm L x 249mm W x 60mm H, 4/Cs RK-34588-26 Mfr # 3175-10
COMPARE
4.5
$235.74CAD / Case of 4
Pyrex 3170-12 3170 Glass Sterilizing Tray, 8L, 4/cs RK-34588-30 Mfr # 3170-12
COMPARE
8
$1,915.41CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat