This product is now discontinued, View Related Products
GK Heller Direct-Drive Mixers; 20 to 2500 rpm (115 VAC, 50/60 Hz)
Maintain constant speed and torque throughout entire speed range
  • X-Y recorder output for charting and calibrating speed and torque
Electronically monitor and record speed and torque. These mixer systems are ideal for polymer chemistry—gather data on viscosity profiles, shear rate and shear stress, starting torque for plastic materials, flocculation points, and more.
All mixers come with a power supply/controller that provides filtered DC power for the motor, changing the voltage as necessary. Maintain constant shaft speed and torque within ±1% of preset values under varying line voltage conditions. Obtain torque readings with a simple flip of a switch.
Permanent magnet, ball-bearing, totally enclosed DC motor has oversized brushes and skewed armature to avoid cogging at low speeds. A fast-acting circuit breaker protects components even if the motor is stalled.
$3,985.52 - $6,659.72CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Motor hp
Availability Pricing
GK Heller HST20N Direct-drive mixer with speed/torque readout; 50 in-oz (max), 115 VAC RK-04406-00 Mfr # 099D HST20N
COMPARE
2500 50 1/8
$5,604.78CAD / Each
GK Heller HST110N Direct-drive mixer with speed/torque readout; 1600 g-cm (max), 115 VAC RK-04406-10 Mfr # 099D HST10N
COMPARE
2500 22.2 1/20
$3,985.52CAD / Each
GK Heller HST30N Direct-drive mixer with speed/torque readout; 100.8 in-oz (max), 115 VAC RK-04406-20 Mfr # 099D HST30N
COMPARE
2500 100.8 1/4
$6,659.72CAD / Each