This product is now discontinued, View Related Products
GE Druck Intrinsically Safe Multifunction Calibrator with Hart Communicator
Modular Calibration and HART Communication System uses three basic components to perform duties formerly requiring a wide range of different instruments
  • Hand held HART communicator and combined multifunction calibrator
  • Powerful device configuration, commissioning and maintenance capabilities
  • Complete library of registered HART device descriptions
  • Free Upgrades and Device Descriptions
  • Integral 24V loop power supply to energize Hart devices and loops
  • Menu selectable 250 loop resistor
  • Measures mA – no need for an ammeter when trimming devices
  • Simultaneously measures and sources the primary variable for sensor trim and device calibration
All components of this modular calibration system are intriniscally safe. The DPI-620-IS multifunction calibrator is an ultra-compact, electrical, frequency and temperature calibrator and HART communicator, that can provide simultaneous measurement and source capabilities with a fully featured HART digital interface.
The high resolution, color touch screen of the calibrator shows only those buttons necessary for the immediate, selected tasks. The buttons have been sized so that they can be operated by a gloved hand, with no need for a stylus. The display can be configured to show up to six reading windows and can show electrical and temperature inputs and outputs, two pressure module readings, an IDOS pressure measurement, and a USB-connected device, as well as a HART summary window or full HART communicator with active mA measurement and actual primary variable readings.
The DPI-620-IS multifunction calibrator now incorporates a fully featured HART® communicator which surpasses market leading communicators in functionality, ease of use and upgrade support. A single handheld device can now configure the full range of HART devices and perform mA trims and sensor calibrations without the need for secondary equipment such as ammeters, calibrators, power supplies and loop resistors. This can significantly reduce equipment inventory, cost of ownership and greatly simplify maintenance activities. The DPI-620-IS contains the latest application software and a complete library of registered HART device descriptions.
$1,364.36 - $14,748.63CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Pressure Pressure Accuracy Pressure Resolution
Availability Pricing
GE Druck DPI 620 IS Intrinsically Safe Advanced Modular Calibrator with HART Communicator RK-50015-01 Mfr # I620IS
COMPARE
$14,748.63CAD / Each
GE Druck DPI 620 IS Intrinsically Safe Advanced Modular Calibrator; HART/Windows CE RK-50015-02 Mfr # I620IS-CE
COMPARE
$10,543.58CAD / Each
GE Druck DPI 620 IS Intrinsically Safe Pressure Module Carrier RK-50015-03 Mfr # MC620-IS
COMPARE
$1,364.36CAD / Each