This product is now discontinued, View Related Products
GE Druck Model DPI 104 Digital Pressure Test Gauges
Use in process monitoring and control and as a test and measurement instrument
  • Standard and Instrinsically-safe versions
  • 0.05% full-scale accuracy
  • Eleven selectable pressure units
  • IDOS compatible and RS232 serial interface
Gauge is a microprossor-controlled precision test gauge that is a reliable and economic solution for a wide range of pressure sensing applications. Gauge features ±0.05% full-scale accuracy, min/max, tare, and alarm functions, analog output, and a pressure switch test function. Gauge can be used as a stand-alone process indicator or as a pressure reference for use with hand pumps. A very unique feature of this guage is the adjustable mounting positions that the user can have. The housing of this gauge rotates 320° around the pressure fitting — faceplate can be rotated in any direction for optimum visibility. In addition to the numeric pressure indication, gauge has a round twenty-segment bar graph for quick quick visual indication. The gauge also has the capability of providing a 1 to 5 VDC output for control and indication.
$1,216.65 - $2,251.34CAD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 30 psia RK-68437-51 Mfr # DPI104-2-30PSIA
COMPARE
Digital Pressure Test Gauge, 0 to 30 psia
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 1000 psia RK-68437-52 Mfr # DPI104-2-1000PSIA
COMPARE
Digital Pressure Test Gauge, 0 to 1000 psia
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 100 psia RK-68437-53 Mfr # DPI104-2-100PSIA
COMPARE
Digital Pressure Test Gauge, 0 to 100 psia
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 10 psig RK-68975-00 Mfr # DPI104-2-10PSIG
COMPARE
Digital Pressure Test Gauge, 0 to 10 psig
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 30 psig RK-68975-02 Mfr # DPI104-2-30PSIG
COMPARE
Digital Pressure Test Gauge, 0 to 30 psig
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 100 psig RK-68975-04 Mfr # DPI104-2-100PSIG
COMPARE
Digital Pressure Test Gauge, 0 to 100 psig
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 300 psig RK-68975-06 Mfr # DPI104-2-300PSIG
COMPARE
Digital Pressure Test Gauge, 0 to 300 psig
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 1000 psig RK-68975-08 Mfr # DPI104-2-1000PSIG
COMPARE
Digital Pressure Test Gauge, 0 to 1000 psig
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 3000 psig RK-68975-10 Mfr # DPI104-2-3000PSIG
COMPARE
Digital Pressure Test Gauge, 0 to 3000 psig
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 5000 psig RK-68975-12 Mfr # DPI104-2-5000PSIG
COMPARE
Digital Pressure Test Gauge, 0 to 5000 psig
$1,217.02CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 10,000 psi, Sealed Gauge RK-68975-14 Mfr # DPI104-2-10000PSISG
COMPARE
Digital Pressure Test Gauge, 0 to 10,000 psi, Sealed Gauge
$1,216.65CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 15,000 psi, Sealed Gauge RK-68975-16 Mfr # DPI104S-4-15000PSI-SG
COMPARE
Digital Pressure Test Gauge, 0 to 15,000 psi, Sealed Gauge
$2,251.34CAD / Each
GE Druck DPI 104 Digital Pressure Test Gauge, 0 to 20,000 psi, Sealed Gauge RK-68975-18 Mfr # DPI104S-4-20000PSI-SG
COMPARE
Digital Pressure Test Gauge, 0 to 20,000 psi, Sealed Gauge
$2,251.34CAD / Each
Showing 1- 13 of 13