This product is now discontinued, View Related Products
Gas Chromatography System for Free and Total Glycerin
Satisfy and perform all of your requirements for ASTM D6584 with this compact, value-priced system
  • Includes powerful PeakSimple™ USB data system — connect GC and your Windows® computer for a complete analytical system
  • Comprehensive display shows the temperature, pressure, and voltage control
  • Ambient to 400°C temperature controllable column oven
  • Electronic Pressure Control (EPC) for all GC system gases, including carrier gas, to ensure user safety
This simple-to-use biodiesel GC system is ideal for the quantitative determination of free and total glycerin in B100 methyl esters by gas chromatography. The calibrated results are displayed directly, eliminating off-line calculations.
Both the carrier gas and the Flame Ionization Detector (FID) combustion gas are controlled by programmable Electronic Pressure Controllers (EPCs). EPCs not only provide rock-solid retention time reproducibility, but allow the carrier gas to be pressure ramped (just as the column oven is temperature ramped) from the built-in PeakSimple data system.
The on-column injector is ideal for 1/8" packed and 0.53-mm wide-bore capillary columns, and is suitable for analyses ranging from methane to heavy, high-boiling hydrocarbons (C44+). The column oven accepts column cage diameters up to four inches, is programmable to 400°C, and recycles quickly with its high-speed cool-down fans.
$1,502.86 - $22,052.23CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Compact BioDiesel Gas Chromatograph with FID for Free/Total Glycerin; 115V
$19,316.14CAD / Each
COMPARE
Compact BioDiesel Gas Chromatograph with FID for Free/Total Glycerin; 230V
$22,052.23CAD / Each
COMPARE
Air compressor option (Built-In) for BioDiesel GC Systems
$1,533.80CAD / Each
COMPARE
Air compressor option (Built-In) for BioDiesel GC System 34009-09
$1,502.86CAD / Each