This product is now discontinued, View Related Products
GammaGuard Sterile Cleanroom Coveralls
Bound seams and tunnelized elastic provide low linting and strong tear resistance
  • Due to the high demand for personal safety products, cancellations and returns will be restricted.
Coveralls feature a sterility assurance of 10-6, low linting, and a more breathable liquid barrier. Special fold design protects against potential contamination during gowning. Feature bound seams and elastic wrists and ankles. Manufactured in an ISO certified facility, clean processed, and individually packaged under cleanroom conditions. Recyclable.
Compliance: ISO Class 4
$434.65CAD / Case of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Color Size
Availability Pricing
International Enviroguard Sterile Cleanroom Coveralls, medium, 25/cs RK-33684-31 Mfr # CE11013CIS-M
COMPARE
White Medium
$434.65CAD / Case of 25
International Enviroguard Sterile Cleanroom Coveralls, large, 25/cs RK-33684-32 Mfr # CE11013CIS-L
COMPARE
White Large
$434.65CAD / Case of 25
International Enviroguard Sterile Cleanroom Coveralls, extra large, 25/cs RK-33684-33 Mfr # CE11013CIS-XL
COMPARE
White X-Large
$434.65CAD / Case of 25
International Enviroguard Sterile Cleanroom Coveralls, double extra large, 25/cs RK-33684-34 Mfr # CE11013CIS-2XL
COMPARE
White XX-Large
$434.65CAD / Case of 25