This product is now discontinued, View Related Products
FW Bell handheld ELF Meter
The 4100 Series Handheld Gaussmeter accurately measures extremely low frequency magnetic fields generated by electrical equipment; ideal for home use
  • Low cost, small, lightweight, and portable
  • High accuracy (< 1%, Model 4190) ± 1% +1 digit
  • 0.1 mG resolution
  • Gauss or Tesla display options
  • Min/Max hold and relative measurement capabilities (4190 only)
  • Selectable X, Y, or Z Axis plus vector summation
  • Waveform and RMS analog output (4190 only)
  • Universal Serial Bus Interface, Remote Operation and USB remote on both 4180 and 4190
  • Unit can be powered from 4 AAA batteries, external DC supply, or USB Bus Power (4190 only)
  • Low battery detection
$1,197.13 - $1,594.21CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Resistance Accuracy Resistance Range Resistance Resolution
Availability Pricing
Pacific Scientific-OECO 4190 Handheld Gaussmeter, Tr-Axis,<1% Accrcy Includes F20 Option RK-20001-34 Mfr # 4190
COMPARE
$1,594.21CAD / Each
Pacific Scientific-OECO 4180 Handheld Tri-Axis Gaussmeter, 2% Accuracy RK-26045-47 Mfr # 4180
COMPARE
$1,197.13CAD / Each