This product is now discontinued, View Related Products
Fluke Vibration Sensing Systems
Monitor vibration measurements to predict machine breakdown
  • Remotely measure vibrations in dangerous or hard-to-reach areas for up to three years!
  • Be alerted to changing conditions to prevent machine failure
  • Compact and lightweight
  • IP67 rated—rugged enough for industrial environments
Make predictive maintenance fast and easy with the Fluke vibration system consisting of a vibration sensor and gateway. Sensor takes continuous overall vibration measurements of acceleration and velocity for a wide range of machines. Long-term vibration monitoring can help detect imbalance, misalignment, looseness, and bearing faults, saving you catastrophic failures and costly downtime. Install gateway within 65 meters from line-of-sight of sensors. Wi-Fi or mobile router/hotspot connectivity and AC power must be available for gateway to work.
Remotely monitor the conditions using the Fluke Connect Condition Monitoring™ (FCCM) software. Set alarms based on user-defined thresholds or Fluke Overall Vibration Severity (FOVS) scale. FCCM software can send alarms to smart devices or computers within seconds of abnormality. View real-time and historical data to take maintenance action.
Connect up to 20 sensors to one gateway. The number of sensors per gateway is determined by two factors: line of sight and distance from sensors. Select from a starter kit or an expansion kit. Starter kit includes a gateway along with four sensors to get you started on monitoring your equipment. Expansion kit includes four sensors. Each kit is available with 1- or 3-year software licenses.
$225.43 - $3,410.90CAD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description Max Vibration Range - Acceleration (g)
Availability Pricing
Fluke FLK- 3561 FC KIT Vibration Sensor Expansion Kit with Software RK-20000-97 Mfr # FLK- 3561 FC KIT
COMPARE
Vibration Sensor Expansion Kit; 1 Yr License 32
$1,757.02CAD / Each
Fluke FLK-3561/3502 FC Vibration Sensor Starter Kit wtih Software RK-20000-98 Mfr # FLK-3561/3502 FC
COMPARE
Vibration Sensor/Gateway Starter Kit; 1 Yr License 32
$1,972.13CAD / Each
FLUKE 3502FC Gateway RK-20000-99 Mfr # FLUKE 3502FC
COMPARE
Gateway
$225.43CAD / Each
Fluke FLK-3561 FC Vibration Sensor Expansion Kit wth Software, 3 Years RK-20001-26 Mfr # FLK-3561 FC 3YR
COMPARE
Vibration Sensor Expansion Kit; 3 Yr License 32
$3,227.46CAD / Each
Fluke FLK-3561/3502 FC Vibration Sensor Starter Kit with Software, 3 Years RK-20001-39 Mfr # FLK-3561/3502 FC 3YR
COMPARE
Vibration Sensor/Gateway Starter Kit; 3 Yr License 32
$3,410.90CAD / Each