Fluke Radiation Survey Meter
Remediate contamination and safety issues while minimally impacting operations
  • Detects skin-dose (beta particle) and deep-dose (gamma) and x-ray radiation
  • Requires no adjustments; simple two button operation
  • Delivers quickly read/correct value through auto ranging capability
  • Automated backlight allows easy visibility inside truck trailers and low-light conditions
  • Rugged sealed case for indoor or outdoor use
  • 30% more accurate than other meters
  • Delivers more than one week of continuous operation from two 9-volt alkaline batteries
  • Proven in use by state and local governmental emergency response professionals, state inspectors, hazmat teams, and nuclear power workers
  • Measures both dose and dose-rate parameters
  • Useful for contamination detection, general radiation area surveys, radiation level monitoring, and hazardous materials assessment
  • Auto-zeroing and auto-ranging
This easy-to-use, handheld radiation detection device measures radiation rate and accumulated dose from beta, gamma, and x-ray radiation sources on irradiated goods, equipment, surfaces, or environments in industrial settings - ensuring worker safety and aiding in federal regulation compliance.
Use this meter everywhere from the receiving dock to warehouse to the shop floor, for scanning vehicles, packages, or even individuals. Unit works at a safe distance, before the user comes into close proximity and encounters unsafe radiation levels, delivering exact information about the extent of contamination allowing you to set up safe perimeters. The programmable flashing LCD and audible alarm notifies the operator of unsafe or out-of-limit conditions, and the intuitive two-button interface makes set up and operation simple - allowing new users to perform surveys with little training.
Applications
Food processing or packaging screening
Transportation goods screening
Imported manufactured goods
Ports
Government and state inspection agencies
Emergency response and hazmat teams (police/fire)
Nuclear power
$1,009.27 - $4,860.71CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Range Description
Availability Pricing
Fluke 190HPS Electronic Corp Carrying Case Single Unit RK-26046-43 Mfr # 190HPS
COMPARE
Electronic Corp Carrying Case Single Unit
$1,009.27CAD / Each
Fluke 481 Radiation Survey/Detection Meter RK-50538-00 Mfr # FLUKE-481
COMPARE
0 to 5 mR/h (8 sec); 0 to 50 mR/h (2.5 sec); 0 to 500 mR/h (2 sec); 0 to 5 R/h (2 sec); 0 to 50 R/h (2 sec) Radiation Survey/Detection Meter
$4,860.71CAD / Each