Fluke Calibration Reference Pressure Gauge/Comparator Kits
Get the best possible performance from your pressure gauges and sensors
  • Easy-to-read 5-1/2 digit gauge display with bar graph
  • Rugged construction for reliable performance
  • Configure, calibrate, and download data with included USB cable
The 2700G series of reference pressure gauges is ideal for calibrating pressure measurement devices and for use as a check standard or as a process gauge with data logging of over 92 kB of memory.
For a complete pressure calibration solution, the model P5500 series of pneumatic and hydraulic test pumps are combined with the 2700G series gauges. Choose between a kit with a single gauge, or a kit with three gauges covering different ranges.
Available for sale in the United States and Canada only
$13,044.72CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Fluke Calibration P5510-2700G-3 Pump and Gauge Kit RK-16115-01 Mfr # P5510-2700G-3
COMPARE
Pump and Gauge Kit
$13,044.72CAD / Each