Fluke Calibration (Hart Scientific) 5611A Series - Silicone-coated secondary thermistor probes
5611A Series – Silicone-coated secondary thermistor probes: With a diameter at the tip of just 1.5 mm ( 1/16”), the 5611A''s tip has the smallest diameter of any of our secondary reference thermistors and can fit nearly anywhere. Its faster response time, flexible sheath, and silicone coating make the 5611A great for use in many applications. However, applications involving silicone oil could damage the thermistor and should be avoided.
Available for sale in the United States and Canada only.
$1,539.69CAD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F)
Availability Pricing
Fluke Calibration 5611A-11-L Bare Thermistor Probe RK-37852-33 Mfr # 5611A-11L
COMPARE
Mini Spade Lug 212
$1,539.69CAD / Each
Fluke Calibration 2739928 Thermistor Probe; 1/16" Dia x 1" L, Bare Wire RK-90450-10 Mfr # 5611A-11-B
COMPARE
Stripped Wire 212
$1,539.69CAD / Each
Fluke Calibration 2739937 Thermistor Probe; 1/16”Dia x 1" L, 5-Pin Din RK-90450-11 Mfr # 5611A-11-D
COMPARE
5-Pin DIN 212
$1,539.69CAD / Each