Fluke Calibration (formerly Hart Scientific) 1523/1524 Reference Thermometers
The first standards thermometers to fit into the palm of your hand.
  • Visual trending and measurement
  • Simultaneously measures two channels
  • Reads PRTs, thermocouples, and thermistors
  • “Plug-and-play” calibration coefficients with SMART LEMO connector
  • Trends data graphically
  • Documents readings and statistics
The 1523 and 1524 are not just basic hand held thermometers, but in fact are highly precise reference thermometers. These rugged thermometers are designed for engineers and technicians that want accuracy and versatility in an easy to use package. Use them with RTD's (PRTs), thermocouples, and thermistors probes to read, document, and visually trend temperature data.
Each unit uses a smart LEMO connector that automatically loads calibration coefficients unique to each probe for easy and accurate plug and play operation. (Only Fluke calibration probes with a "P" style Info-Con connector will fit this meter)
Two models are available distinguished by the number of channels they measure and data logging capability. Includes a free software program that makes it easy to download logged data and program new calibration coefficients.
Available for sale in the United States and Canada only.
$3,916.27 - $4,678.01CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Meter Measurement Type Number Of Probe Channels
Availability Pricing
Fluke Calibration 1523 Handheld Thermometer Model RK-93202-50 Mfr # 1523-156
COMPARE
3182 Multi-Parameter 1
$3,916.27CAD / Each
Fluke Calibration 1524 Handheld Thermometer with Datalogger Model RK-93202-51 Mfr # 1524-156
COMPARE
3182 Multi-Parameter 2
$4,678.01CAD / Each