Fluke Calibration 6109A and 7109A Portable Calibration Baths

Increase efficiency and reduce production downtime
  • Calibrate up to four sanitary temperature sensors at a time—not just one
  • Wide temperature range covers most clean-process operations
  • Display accuracy of ±0.1°C provides 4:1 test uncertainty ratio (TUR) for critical applications
  • Cleanroom compatible—stainless steel case is rustproof and withstands sterilizing chemicals
  • Easy to transport, even with one hand
These portable baths increase efficiency and reduce downtime with twice the accuracy of other calibrators in its class. Calibrate up to four Tri-Clamp® sanitary temperature sensors at a time, more for smaller or flangeless sensors, and bring the bath to your process to save production downtime.
Choose from two temperature ranges to cover your application needs. Models with process electronics include accessories for connecting an external reference probe.
$16,185.19 - $22,587.07CAD / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Power (VAC) Description
Availability Pricing
Fluke Calibration 6109A-156 Portable Bath, 35 to 250°C, 115 VAC RK-16108-44 Mfr # 6109A-156
COMPARE
482 115 Portable Calibration Bath, 35 to 250°C, 115 VAC
$16,185.19CAD / Each
Fluke Calibration 6109A-P-156 Portable Bath, 35 to 250°C, with Process Electronics, 115 VAC RK-16108-45 Mfr # 6109A-P-156
COMPARE
482 115 Portable Calibration Bath, 35 to 250°C, with Process Electronics, 115 VAC
$19,419.28CAD / Each
Fluke Calibration 7109A-156 Portable Bath, -25 to 140°C, 115 VAC RK-16108-46 Mfr # 7109A-156
COMPARE
284 115 Portable Calibration Bath, -25 to 140°C, with Process Electronics, 115 VAC
$19,367.71CAD / Each
Fluke Calibration 7109A-P-156 Portable Bath, -25 to 140°C, with Process Electronics, 115 VAC RK-16108-47 Mfr # 7109A-P-156
COMPARE
284 115 Portable Calibration Bath, -25 to 140°C, 115 VAC
$22,587.07CAD / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At800-363-5900
Live chat