This product is now discontinued, View Related Products
Fluke 1760 Three Phase Power Quality Recorder
Class A compliance for the most demanding power quality tests
  • Fully Clas A compliant
  • GPS time synchronization
  • Flexible and fully configurable thresholds and scale factors
  • Uninterrupted power supply (40 minutes)
  • 10 MHz, 6000 Vpk waveform capture
  • 2 GB data memory
  • Comprehensive software included
  • Plug and play rugged field design
The Fluke 1760 Three-Phase Power Quality Recorder is fully compliant to IEC 61000-4-30 Class-A, for advanced power quality analysis and consistent compliance testing. Designed for analysis of utility and industrial power distribution systems, in medium- and low-voltage networks, this power quality monitor provides the flexibility to customize thresholds, algorithms, and measurement selections. The 1760 power recorder captures the most comprehensive details on user-selected parameters.
Sold in the United States and Canada only
$26,521.01 - $44,785.15CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) DC Max (Volts) Description
Availability Pricing
Fluke 1760 BASIC Power Quality Recorder, No V or I Probes, No Trans RK-20035-75 Mfr # FLUKE-1760 BASIC
COMPARE
6000 6000 Power Quality Recorder, No V or I Probes, No Trans
$26,521.01CAD / Each
Fluke 1760 TR US Power Quality Recorder with Trans RK-20035-76 Mfr # 20430
COMPARE
6000 6000 Power Quality Recorder with Trans
$44,785.15CAD / Each
Fluke 1760 US Power Quality Recorder, no Trans RK-20035-77 Mfr # FLUKE-1760 US
COMPARE
6000 6000 Power Quality Recorder, no Trans
$31,309.52CAD / Each
Fluke 1760 TR Basic Power Quality Recorder, with Trans, no V or I Probes RK-20035-78 Mfr # 20430
COMPARE
6000 6000 Power Quality Recorder, with Trans, no V or I Probes
$39,856.66CAD / Each