Fluke 1732 and 1734 Three-Phase Electrical Energy Loggers
Discover inefficiencies and optimize your energy bill
  • Automatically capture and log voltage, current, power, power factor, energy and associated values
  • Conserve instrument battery—power instrument directly from the measured circuit
  • Measure all three phases—3 flexible current probes included
  • Perform in-the-field analysis and data checks on the touch screen, graphical display
  • Store more than 20 separate logging sessions
  • CAT IV-600 V/ CAT III-1000 V safety rating
  • Ideal for load studies, energy assessments, and energy surveys
Find when and where energy in your facility is being consumed—from the service entrance to individual circuits. These energy loggers introduce a new simplicity to discovering sources of electrical energy waste. View data locally on the instrument, through Fluke Connect® mobile app and desktop software, or through your facilities’ Wi-Fi infrastructure. All models are Fluke Connect® compatible, model 20000-93 includes the Wi-Fi/BLE adapter to enable it. Download and analyze every detail of energy consumption with automated reporting. Every session detail is stored so you don't lose measurement trends—automatically.
$7,906.20CAD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Max AC Voltage (Voltage) Description
Availability Pricing
Fluke 1734/EUS Energy Logger, Advanced with FlukeConnect®, with Flex Probes RK-20000-93 Mfr # FLUKE-1734/EUS
COMPARE
1000 Energy Logger, Advanced with FlukeConnect®; EU/US/UK
$7,906.20CAD / Each