FlowStat Sight Flow Transmitters
Output signal for local and remote indication
These sight flow sensors are designed for low-cost applications where visual indication and sensing output signals are required—available with 4 to 20 mA, pulse, relay, or 0 to 5 VDC output. Durable polypropylene body is compatible with a variety of fluids. Sensors offer a flow measuring accuracy of ±2% full-scale, and are capable of handling pressures up to 150 psig and temperatures up to 150°F (66°C).
$234.27 - $450.86CAD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Min Flow Rate (GPM) Output
Availability Pricing
AW-Lake CCB Sight Flow Transmitter, 4-20 mA, 0.5-5 GPM, 12-35 VDC RK-32825-00 Mfr # CCB-05-B-ES
COMPARE
4 to 20 mA
$450.86CAD / Each
AW-Lake CCB Sight Flow Transmitter, 4-20 mA, 1-10 GPM, 12-35 VDC RK-32825-02 Mfr # CCB-10-B-ES
COMPARE
4 to 20 mA
$450.86CAD / Each
AW-Lake CCB Sight Flow Transmitter, 4-20 mA, 1.5-15 GPM, 12-35 VDC RK-32825-04 Mfr # CCB-15-B-ES
COMPARE
4 to 20 mA
$450.86CAD / Each
AW-Lake PCB Sight Flow Transmitter, Pulse, 0.5-5 GPM, 15-24 VDC RK-32825-06 Mfr # PCB-05-B-ES
COMPARE
Pulse
$234.27CAD / Each
AW-Lake PCB Sight Flow Transmitter, Pulse, 1.5-15 GPM, 15-24 VDC RK-32825-10 Mfr # PCB-15-B-ES
COMPARE
Pulse
$234.27CAD / Each
AW-Lake RCB Sight Flow Transmitter, Relay, 0.5-5 GPM, 12-35 VDC RK-32825-12 Mfr # RCB-05-B-ES
COMPARE
Relay
$360.98CAD / Each
AW-Lake RCB Sight Flow Transmitter, Relay, 1-10 GPM, 12-35 VDC RK-32825-14 Mfr # RCB-10-B-ES
COMPARE
Relay
$372.77CAD / Each
AW-Lake VCB Sight Flow Transmitter, 0-5 VDC Output, 0.5 to 5 GPM, 12-35 VDC RK-32825-18 Mfr # VCB-05-B-ES
COMPARE
0 to 5 VDC
$421.39CAD / Each
AW-Lake VCB Sight Flow Transmitter, 0-5 VDC Output, 1-10 GPM, 12-35 VDC RK-32825-20 Mfr # VCB-10-B-ES
COMPARE
0 to 5 VDC
$421.39CAD / Each
AW-Lake VCB Sight Flow Transmitter, 0-5 VDC Output, 1.5-15 GPM, 12-35 VDC RK-32825-22 Mfr # VCB-15-B-ES
COMPARE
0 to 5 VDC
$421.39CAD / Each
Showing 1- 10 of 10