This product is now discontinued, View Related Products
Flow-Through Heater
Use this flow-through heater to pump and heat large quantities of liquids. It features a single-turn potentionmeter for easy temperature setting, a high-temperature cutoff at 203ºF (95ºC), and a no-flow heater cutoff to prevent heating without flow. Seal the reservoir to control tanks or closed loops systems.
The built-in, centifungal-style submersible pump must be primed before use. Wetted parts consist of polypropylene body, nylon impeller, and stainless steel shaft. Includes one 2-ft sections of 1/2" ID vinyl tubing and 6-ft power cord; 120 VAC model includes three-prong plug.
$1,850.58 - $2,638.84CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Power (VAC) Power (watts)
Availability Pricing
PolyScience 040300 Flow-Through Heater, 750 Watts, 120 V, 60 Hz RK-01285-80 Mfr # 40300
COMPARE
158 120 750
$2,638.84CAD / Each
PolyScience 040301 Flow-Through Heater, 750 Watts, 240 V, 50 Hz RK-01285-85 Mfr # 040301
COMPARE
158 240 750
$1,850.58CAD / Each