Flow Monitors and Accessories
Monitor both flow rate and flow from pulse output flowmeters in any engineering unit with a push of a button. Programmable, compact, and easy-to-use. Input square wave, sine, or sawtooth frequency. Back-up totalizer lock-out feature prevents accidental erasure. High-speed microprocessor ensures fast and accurate reporting — includes a 4 to 20 mA output for recording or datalogging. Model 32928-76 has two channels to monitor dual flows and display them separately, as a sum, as a difference, or as a ratio.
$1,052.00 - $1,222.91CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Output
Availability Pricing
AW-Lake FEM-03 Gear Flow Sensor/Transmitter Accessory, Flow Monitor RK-32928-75 Mfr # MX9-SX-XX
COMPARE
$1,052.00CAD / Each
AW-Lake FEM-03A Gear Flow Sensor/Transmitter Accessory, Flow Monitor w/ 4-20 mA Output RK-32928-76 Mfr # MX9-S4-XX
COMPARE
$1,222.91CAD / Each