This product is now discontinued, View Related Products
FLIR Vibrating Noncontact Voltage Detectors with Flashlight
Vibration and flashing LED alarms indicate presence of voltage
  • Tip light permits operation in dimly lit areas
  • Withstands a drop from 10 ft (3 m)
  • Conserve battery life with auto power-off
  • Execute tasks efficiently with built-in LED flashlight
  • CAT IV safety rating
The VP-series of noncontact voltage detectors quickly and easily alert you of live and neutral wires in sockets and junction boxes with vibration feedback and flashing red LED alarms. Low-profile probe tip allows you to safely place the sensor close to electrical sources to detect voltages. The ergonomic design with double molded slip-resistant grip and an anti-roll case reduces the chance of dropping the unit. The detectors also include high and low sensitivity ranges, a bright LED flashlight, probe tip light, low-battery indicator, auto power off, and a CAT IV-100V safety rating.
$48.61 - $51.55CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
AC Max (Volts) AC Min (Volts) Description
Availability Pricing
Flir VP50 Noncontact Voltage Detector with Flashlight; 24-1000 VAC RK-20046-23 Mfr # VP50
COMPARE
1000 24 Noncontact Voltage Detector with flashlight detects voltage on the latest tamperproof outlets; ; 24-1000 V
$51.55CAD / Each
Flir VP40 Noncontact Voltage Detector with Flashlight; 24-1000 VAC RK-39753-33 Mfr # VP40
COMPARE
1000 90 Noncontact AC Voltage Detector with Flashlight; 90-1000 VAC
$48.61CAD / Each