This product is now discontinued, View Related Products
FLIR® True RMS Clamp Meters with Worklights
Integrated LED worklights make cabinet measurements easier
  • Compact jaw and slim design fit in narrow workspaces and pockets
  • Light up the dark — worklights are built into the base of the jaws
  • Get accurate readings with features like True RMS, LoZ, smart diode, and an input for test leads
Compact, rugged design fits perfectly in your hand and is designed to withstand the most challenging industrial environments—including low-light, crowded panels. The narrow jaw with integrated worklights makes working in cramped, dark cabinets safer and easier. Meters feature a large backlit LCD, Min/Max and Hold functions, and auto-power off (may be disabled).
$220.99 - $428.76CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Flir CM72 FLIR True RMS Clamp Meter with Worklights; 600 V RK-20046-72 Mfr # CM72
COMPARE
FLIR True RMS Clamp Meter with Worklights; 600 V
$220.99CAD / Each
Flir CM74 FLIR True RMS Clamp Meter with Worklights; 1000 V RK-20046-74 Mfr # CM74
COMPARE
FLIR True RMS Clamp Meter with Worklights; 1000 V
$428.76CAD / Each