FLIR® TG-Series Diagnostic Thermal Cameras
See beyond the limits of a single-spot IR thermometer with improved image resolution
  • IP54 enclosure protects from dirt, dust, and oil
  • Accurate targeting through the help of the laser pointer and cross-hair display ensures precise temperature measurement
  • Visible spectrum 2M pixel digital camera provides excellent picture quality
The lightweight, compact, and rugged TG-series infrared thermometers are designed to help you quickly and accurately diagnose hot and cold trouble spots from a safe distance. Easily record before and after thermal images to confirm problems have been corrected. Images are clearer with MSX® two-camera technology that adds crisp visual details. The intuitive icon menu system is simple to operate. Other features include selectable color palette, emissivity adjustment, selectable temperature scale, and image hold. The TG267 and TG297 support Bluetooth® real-time remote temperature monitoring and transmission to mobile devices. There are three models to choose from depending on your needs.
FLIR TG165-X measures temperatures from -13 to 572°F (-25 to 300°C) with a distance-to-spot ratio of 24:1.
FLIR TG267 can accommodate a Type K thermocouple for contact measurement readings up to 500°F (260°C). The IR camera measures a wider range of temperatures from -13 to 716°F (-25 to 380°C) with a distance-to-spot ratio of 24:1.
FLIR TG297 is similar to the TG267 but features a high-temperature filter which measures temperatures up to 1886°F (1030°C) with a distance-to-spot ratio of 30:1.
$823.63 - $1,648.72CAD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Detector Size Max Temperature (° F) Pixels / Thermal Resolution
Availability Pricing
Flir TG267 Diagnostic Thermal Camera, 160 x 120 Resolution RK-39757-48 Mfr # TG267
COMPARE
160 x 120 716 19200
$823.63CAD / Each
Flir TG297 Diagnostic Thermal Camera, 160 x 120 Resolution, High Temperature RK-39757-49 Mfr # TG297
COMPARE
160 x 120 1886 19200
$1,648.72CAD / Each