This product is now discontinued, View Related Products
FLIR FC-Series Automation Thermal Cameras
Get optimal detection with long-term reliability
  • On-board video analytics with ability to classify human or vehicle intrusions
  • Multiple fields of view and resolution options
  • Numerous alarm notifications
The fixed network thermal cameras feature up to four on-board temperature measurement tools and multiple safety alarm notifications. When a monitored temperature of a specific area has been reached or exceeded, a notification goes out by email, digital output or VMS alarm. Cameras can be calibrated for fire detection, safety, and thermal monitoring of substations, waste disposal, and equipment. A powerful on-board analytics is capable of classifying human or vehicle intrusions. Flexible integration tools allow temperature data and alarms to be integrated into a wide variety of external monitoring and control systems. The thermal cameras can be installed with an analog or digital (IP) video output or both. Analog video requires a connection to a video monitor or an analog video matrix switch. A rugged constructed enclosure protects the cameras from dust, water, and is submersible up to one meter (3 1/4 ft). They are also shock, vibration, and corrosion-resistant. The cameras are intended for outdoor security applications and can be install in a fixed location using wall mount (not included) or on a pan/tilt mechanism.
$5,355.77 - $10,962.02CAD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Flir FC-324R Fixed Network Thermal Camera, 320 x 240 Array RK-39756-99 Mfr # FC-324 R-19MM NTSC
COMPARE
Fixed Network Thermal Camera, 320 x 240 Array
$5,355.77CAD / Each
Flir FC-334R Fixed Network Thermal Camera, 320 x 240 Array RK-39757-00 Mfr # FC-334 R-13MM NTSC
COMPARE
Fixed Network Thermal Camera, 320 x 240 Array
$5,355.77CAD / Each
Flir FC-632R Fixed Network Thermal Camera, 640 x 480 Array RK-39757-01 Mfr # FC-632 R-19MM NTSC
COMPARE
Fixed Network Thermal Camera, 640 x 480 Array
$10,962.02CAD / Each
Flir FC-645R Fixed Network Thermal Camera, 640 x 480 Array RK-39757-02 Mfr # FC-645 R-13MM NTSC
COMPARE
Fixed Network Thermal Camera, 640 x 480 Array
$10,962.02CAD / Each